Learn More
Meso Scale Discovery UPLEX HUM LAG3 ASY384well
SDP
Catalog No. 5000081046
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human LAG3 protein in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a typical lower limit of detection of 19 pg/mL and includes reagents for one standard curve and three samples This quantitative immunoassay provides sensitive reproducible and rapid quantification of LAG3 levels
Catalog No. 50-000-81046 Supplier Meso Scale Discovery Supplier No. K251Q7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LAG3 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human LAG3 protein in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a typical lower limit of detection of 19 pg/mL and includes reagents for one standard curve and three samples This quantitative immunoassay provides sensitive reproducible and rapid quantification of LAG3 levels