Learn More
Meso Scale Discovery UPLEX HUM LAG3 ASYQP 25 PL
SDP
Catalog No. 5000081668
Encompass
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay for detecting and quantifying LAG3 and other biomarkers in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for LAG3
Catalog No. 50-000-81668 Supplier Meso Scale Discovery Supplier No. K151Q7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LAG3 Assay Kit is a multiplex immunoassay for detecting and quantifying LAG3 and other biomarkers in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL for LAG3