Learn More
Meso Scale Discovery UPLEX HUM Leptin ASY384well
SDP
Catalog No. 5000078461
Encompass
The Meso Scale Discovery U-PLEX Human Leptin 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human leptin in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity broad dynamic range and low sample volume requirements The kit includes a 384-well MULTI-ARRAY plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The assay has a typical sensitivity of 0 05 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78461 Supplier Meso Scale Discovery Supplier No. K2515ZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Leptin 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human leptin in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity broad dynamic range and low sample volume requirements The kit includes a 384-well MULTI-ARRAY plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The assay has a typical sensitivity of 0 05 pg/mL and an upper limit of quantification of 10 000 pg/mL