Learn More
Meso Scale Discovery UPLEX HUM Leptin ASY384well
SDP
Catalog No. 5000080783
Encompass
The Meso Scale Discovery U-PLEX Human Leptin 384-Well Assay Kit is designed for the quantitative measurement of leptin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time-to-result This 384-well assay kit includes a plate coated with capture antibody detection antibody conjugated to an ECL label calibrator and buffer The assay can detect leptin concentrations from 0 06-1500 pg/mL and samples require just 2 L per well
Catalog No. 50-000-80783 Supplier Meso Scale Discovery Supplier No. K2515ZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Leptin 384-Well Assay Kit is designed for the quantitative measurement of leptin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time-to-result This 384-well assay kit includes a plate coated with capture antibody detection antibody conjugated to an ECL label calibrator and buffer The assay can detect leptin concentrations from 0 06-1500 pg/mL and samples require just 2 L per well