Learn More
Meso Scale Discovery UPLEX HUM LH ASY384well
SDP
Catalog No. 5000080784
Encompass
The Meso Scale Discovery U-PLEX Human LH 384-Well Assay is an electrochemiluminescent immunoassay for measuring luteinizing hormone (LH) in human serum plasma and culture media It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The assay has a dynamic range of 0 04 - 1000 mIU/mL and requires 25 L of sample It supports research in reproductive health fertility and endocrinology The kit includes a 384-well plate with capture antibody detection antibody calibrator and buffer for 192 samples in duplicate
Catalog No. 50-000-80784 Supplier Meso Scale Discovery Supplier No. K2516FK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human LH 384-Well Assay is an electrochemiluminescent immunoassay for measuring luteinizing hormone (LH) in human serum plasma and culture media It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The assay has a dynamic range of 0 04 - 1000 mIU/mL and requires 25 L of sample It supports research in reproductive health fertility and endocrinology The kit includes a 384-well plate with capture antibody detection antibody calibrator and buffer for 192 samples in duplicate