Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASY384
SDP
Catalog No. 5000080746
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of human TNFSF14 in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the rapid and efficient analysis of TNFSF14 levels with as little as 25 L of sample The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay
Catalog No. 50-000-80746 Supplier Meso Scale Discovery Supplier No. K251AHCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of human TNFSF14 in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the rapid and efficient analysis of TNFSF14 levels with as little as 25 L of sample The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay