Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASY384
SDP
Catalog No. 5000081119
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a 384-well multiplex immunoassay kit for quantifying human TNFSF14 in cell culture supernatants serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs TNFSF14 also known as LIGHT or CD258 is crucial for immune cell activation The kit includes a plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and quantifying TNFSF14
Catalog No. 50-000-81119 Supplier Meso Scale Discovery Supplier No. K251AHCK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a 384-well multiplex immunoassay kit for quantifying human TNFSF14 in cell culture supernatants serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs TNFSF14 also known as LIGHT or CD258 is crucial for immune cell activation The kit includes a plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and quantifying TNFSF14