Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASYQP
SDP
Catalog No. 5000078863
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a multiplex immunoassay that measures TNFSF14 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms The U-PLEX format allows simultaneous measurement of TNFSF14 with other biomarkers This assay could be useful for studying TNFSF14 biology and immune function
Catalog No. 50-000-78863 Supplier Meso Scale Discovery Supplier No. K151AHCK22
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a multiplex immunoassay that measures TNFSF14 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms The U-PLEX format allows simultaneous measurement of TNFSF14 with other biomarkers This assay could be useful for studying TNFSF14 biology and immune function