Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASYQP
SDP
Catalog No. 5000081626
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 Assay is a multiplex immunoassay for measuring TNFSF14 levels in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L or less it is compatible with MESO QuickPlex SQ 120 SQ 120 Pro and SECTOR S 600 It measures TNFSF14 from 0 06 to 2500 pg/mL with 10% CV precision enabling rapid reproducible quantification
Catalog No. 50-000-81626 Supplier Meso Scale Discovery Supplier No. K151AHCK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 Assay is a multiplex immunoassay for measuring TNFSF14 levels in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L or less it is compatible with MESO QuickPlex SQ 120 SQ 120 Pro and SECTOR S 600 It measures TNFSF14 from 0 06 to 2500 pg/mL with 10% CV precision enabling rapid reproducible quantification