Learn More
Meso Scale Discovery UPLEX HUM MCP1 ASYQP 25 PL
SDP
Catalog No. 5000080392
Encompass
The U-PLEX Human MCP-1 Assay (K151UGK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure Monocyte Chemoattractant Protein-1 (MCP-1/CCL2) levels in human biological samples This multiplex-ready assay utilizes Meso Scale Discovery s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying inflammation immune response and cardiovascular disease biomarkers
Catalog No. 50-000-80392 Supplier Meso Scale Discovery Supplier No. K151UGK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MCP-1 Assay (K151UGK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure Monocyte Chemoattractant Protein-1 (MCP-1/CCL2) levels in human biological samples This multiplex-ready assay utilizes Meso Scale Discovery s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying inflammation immune response and cardiovascular disease biomarkers