Learn More
Meso Scale Discovery UPLEX HUM MCP2 ASY384well
SDP
Catalog No. 5000080515
Encompass
The Meso Scale Discovery U-PLEX Human MCP-2 (384-Well) Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human monocyte chemoattractant protein 2 (MCP-2) in serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL Results are obtained in as little as 3 5 hours using Meso Scale Discovery instrumentation This assay allows high-throughput quantification of MCP-2 levels
Catalog No. 50-000-80515 Supplier Meso Scale Discovery Supplier No. K251XHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MCP-2 (384-Well) Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human monocyte chemoattractant protein 2 (MCP-2) in serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL Results are obtained in as little as 3 5 hours using Meso Scale Discovery instrumentation This assay allows high-throughput quantification of MCP-2 levels