Learn More
Meso Scale Discovery UPLEX HUM MCSF ASY384well
SDP
Catalog No. 5000081133
Encompass
The Meso Scale Discovery MULTI-ARRAY M-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants The assay utilizes MSD s patented MULTI-ARRAY technology and provides high sensitivity a wide dynamic range and requires a low sample volume The lower limit of detection is 1 3 pg/mL The assay can be run on MSD s SECTOR Imager 6000 for high throughput screening This quantitative immunoassay allows sensitive reproducible and rapid measurement of M-CSF
Catalog No. 50-000-81133 Supplier Meso Scale Discovery Supplier No. K251XRK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MULTI-ARRAY M-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of macrophage colony-stimulating factor (M-CSF) in human serum plasma and cell culture supernatants The assay utilizes MSD s patented MULTI-ARRAY technology and provides high sensitivity a wide dynamic range and requires a low sample volume The lower limit of detection is 1 3 pg/mL The assay can be run on MSD s SECTOR Imager 6000 for high throughput screening This quantitative immunoassay allows sensitive reproducible and rapid measurement of M-CSF