Learn More
Meso Scale Discovery UPLEX HUM MCSF ASYQP 25 PL
SDP
Catalog No. 5000081814
Encompass
The U-PLEX Human M-CSF Assay (K151XRK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of Macrophage Colony-Stimulating Factor (M-CSF) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable M-CSF quantification in serum plasma and cell culture supernatants for research applications
Catalog No. 50-000-81814 Supplier Meso Scale Discovery Supplier No. K151XRK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human M-CSF Assay (K151XRK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of Macrophage Colony-Stimulating Factor (M-CSF) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable M-CSF quantification in serum plasma and cell culture supernatants for research applications