Learn More
Meso Scale Discovery UPLEX HUM MDC ASY384well
SDP
Catalog No. 5000080616
Encompass
The U-PLEX Human MDC Assay from Meso Scale Diagnostics is a multiplex immunoassay panel for measuring 10 human myeloid-related proteins simultaneously It uses electrochemiluminescent detection on a proprietary multi-array plate The panel includes IFN-alpha2a IL-1beta IL-6 IL-8 IL-10 IL-12p70 IL-23 IL-27 TNF-alpha and VEGF-A It requires 50 L of sample per well with a total assay time of about 3 5 hours The U-PLEX platform offers high sensitivity broad dynamic range and low sample volume suitable for biomarker research in immunology inflammation and oncology
Catalog No. 50-000-80616 Supplier Meso Scale Discovery Supplier No. K251UIK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MDC Assay from Meso Scale Diagnostics is a multiplex immunoassay panel for measuring 10 human myeloid-related proteins simultaneously It uses electrochemiluminescent detection on a proprietary multi-array plate The panel includes IFN-alpha2a IL-1beta IL-6 IL-8 IL-10 IL-12p70 IL-23 IL-27 TNF-alpha and VEGF-A It requires 50 L of sample per well with a total assay time of about 3 5 hours The U-PLEX platform offers high sensitivity broad dynamic range and low sample volume suitable for biomarker research in immunology inflammation and oncology