Learn More
Meso Scale Discovery UPLEX HUM MIG ASY384well
SDP
Catalog No. 5000081107
Encompass
The U-PLEX Human MIG Assay 384-well Kit from Meso Scale Discovery is a 384-well multiplex immunoassay kit for the quantitative measurement of human MIG in cell culture supernatants serum plasma and other biological fluids The kit contains a 384-well plate coated with capture antibodies for MIG detection antibodies calibrators and buffers The assay uses electrochemiluminescence for high sensitivity and broad dynamic range Results are obtained in as little as 3 5 hours using an MSD instrument The kit can quantify MIG concentrations between 0 05-1000 pg/mL
Catalog No. 50-000-81107 Supplier Meso Scale Discovery Supplier No. K251AGYK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MIG Assay 384-well Kit from Meso Scale Discovery is a 384-well multiplex immunoassay kit for the quantitative measurement of human MIG in cell culture supernatants serum plasma and other biological fluids The kit contains a 384-well plate coated with capture antibodies for MIG detection antibodies calibrators and buffers The assay uses electrochemiluminescence for high sensitivity and broad dynamic range Results are obtained in as little as 3 5 hours using an MSD instrument The kit can quantify MIG concentrations between 0 05-1000 pg/mL