Learn More
Meso Scale Discovery UPLEX HUM MIP1 ASY 25 PL
SDP
Catalog No. 5000080532
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1alpha Assay Kit is designed for the quantitative measurement of human MIP-1alpha in serum plasma cell culture supernatants cell and tissue lysates and other biological samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid time to results The assay requires a small sample volume and minimal hands-on time It includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and vials of lyophilized U-PLEX Calibrator and Controls The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80532 Supplier Meso Scale Discovery Supplier No. K151UJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-1alpha Assay Kit is designed for the quantitative measurement of human MIP-1alpha in serum plasma cell culture supernatants cell and tissue lysates and other biological samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid time to results The assay requires a small sample volume and minimal hands-on time It includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and vials of lyophilized U-PLEX Calibrator and Controls The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL