Learn More
Meso Scale Discovery UPLEX HUM MIP1 ASY384well
SDP
Catalog No. 5000080704
Encompass
The Meso Scale Discovery Human MIP-1 alpha U-PLEX Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of MIP-1 alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and wide dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It provides reproducible and reliable results for human MIP-1 alpha with a lower limit of detection of 0 04 pg/mL This assay is for research use only and not for use in diagnostic procedures
Catalog No. 50-000-80704 Supplier Meso Scale Discovery Supplier No. K251UJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human MIP-1 alpha U-PLEX Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of MIP-1 alpha in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and wide dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It provides reproducible and reliable results for human MIP-1 alpha with a lower limit of detection of 0 04 pg/mL This assay is for research use only and not for use in diagnostic procedures