Learn More
Meso Scale Discovery UPLEX HUM MIP1 ASY384well
SDP
Catalog No. 5000081082
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1alpha 384-Well Assay Kit is a multiplex immunoassay for measuring human MIP-1alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and optimized diluents It enables simultaneous measurement of MIP-1alpha and other biomarkers ensuring high performance and reliability in biomarker quantification
Catalog No. 50-000-81082 Supplier Meso Scale Discovery Supplier No. K251UJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-1alpha 384-Well Assay Kit is a multiplex immunoassay for measuring human MIP-1alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and optimized diluents It enables simultaneous measurement of MIP-1alpha and other biomarkers ensuring high performance and reliability in biomarker quantification