Learn More
Meso Scale Discovery UPLEX HUM MIP1B ASY 25 PL
SDP
Catalog No. 5000080393
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1beta Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MIP-1beta in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent It allows sensitive reproducible and rapid quantification of the MIP-1beta biomarker
Catalog No. 50-000-80393 Supplier Meso Scale Discovery Supplier No. K151UKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-1beta Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MIP-1beta in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent It allows sensitive reproducible and rapid quantification of the MIP-1beta biomarker