Learn More
Meso Scale Discovery UPLEX HUM MIP1B ASY384well
SDP
Catalog No. 5000080518
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-1beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent It allows simultaneous detection of human MIP-1beta along with other biomarkers in the U-PLEX platform The kit has high performance and is ideal for biomarker research and drug development applications
Catalog No. 50-000-80518 Supplier Meso Scale Discovery Supplier No. K251UKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-1beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-1beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent It allows simultaneous detection of human MIP-1beta along with other biomarkers in the U-PLEX platform The kit has high performance and is ideal for biomarker research and drug development applications