Learn More
Meso Scale Discovery UPLEX HUM MIP1B ASYQP 25 PL
SDP
Catalog No. 5000079719
Encompass
The U-PLEX Human MIP-1 Assay (K151UKK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of MIP-1 (Macrophage Inflammatory Protein-1 beta) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications
Catalog No. 50-000-79719 Supplier Meso Scale Discovery Supplier No. K151UKK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human MIP-1 Assay (K151UKK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of MIP-1 (Macrophage Inflammatory Protein-1 beta) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications