Learn More
Meso Scale Discovery UPLEX HUM MIP3 ASY384well
SDP
Catalog No. 5000080621
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well plate assay allows for the simultaneous measurement of MIP-3alpha along with other biomarkers It only requires 10-25 L of sample per well and the entire workflow can be completed in around 3 hours With minimal hands-on time required this assay provides a rapid and efficient way to quantify MIP-3alpha levels across large numbers of samples
Catalog No. 50-000-80621 Supplier Meso Scale Discovery Supplier No. K251UZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well plate assay allows for the simultaneous measurement of MIP-3alpha along with other biomarkers It only requires 10-25 L of sample per well and the entire workflow can be completed in around 3 hours With minimal hands-on time required this assay provides a rapid and efficient way to quantify MIP-3alpha levels across large numbers of samples