Learn More
Meso Scale Discovery UPLEX HUM MIP3 ASY384well
SDP
Catalog No. 5000081125
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay for measuring human MIP-3alpha in various biological fluids It uses electrochemiluminescence detection for high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies labeled detection antibodies and calibrators for a standard curve It enables simultaneous measurement of MIP-3alpha and other biomarkers offering high performance and reliability for biomarker research and drug discovery
Catalog No. 50-000-81125 Supplier Meso Scale Discovery Supplier No. K251UZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay for measuring human MIP-3alpha in various biological fluids It uses electrochemiluminescence detection for high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies labeled detection antibodies and calibrators for a standard curve It enables simultaneous measurement of MIP-3alpha and other biomarkers offering high performance and reliability for biomarker research and drug discovery