Learn More
Meso Scale Discovery UPLEX HUM MIP3B ASY 25 PL
SDP
Catalog No. 5000078766
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3beta Assay Kit quantifies human Macrophage Inflammatory Protein 3 beta (MIP-3beta) in serum plasma and cell culture supernatants using electrochemiluminescent detection It features a plate with capture antibodies and a U-PLEX linker for analyte binding Detection antibodies with an electrochemiluminescent label bind the analyte Voltage applied to plate electrodes in MSD Read Buffer T causes light emission measured by MSD instruments The kit includes a 96-well plate detection antibody calibrator and assay diluent with sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-78766 Supplier Meso Scale Discovery Supplier No. K151VAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-3beta Assay Kit quantifies human Macrophage Inflammatory Protein 3 beta (MIP-3beta) in serum plasma and cell culture supernatants using electrochemiluminescent detection It features a plate with capture antibodies and a U-PLEX linker for analyte binding Detection antibodies with an electrochemiluminescent label bind the analyte Voltage applied to plate electrodes in MSD Read Buffer T causes light emission measured by MSD instruments The kit includes a 96-well plate detection antibody calibrator and assay diluent with sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL