Learn More
Meso Scale Discovery UPLEX HUM MIP3B ASY384well
SDP
Catalog No. 5000080171
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma cell culture supernatants cell and tissue lysates and other biological fluids This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption It is available in a 384-well plate format and includes all necessary reagents and plates to run the assay This quantitative immunoassay allows simultaneous detection of multiple biomarkers and analytes from a small sample volume
Catalog No. 50-000-80171 Supplier Meso Scale Discovery Supplier No. K251VAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-3beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma cell culture supernatants cell and tissue lysates and other biological fluids This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption It is available in a 384-well plate format and includes all necessary reagents and plates to run the assay This quantitative immunoassay allows simultaneous detection of multiple biomarkers and analytes from a small sample volume