Learn More
Meso Scale Discovery UPLEX HUM MIP3B ASY384well
SDP
Catalog No. 5000080666
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer It can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-80666 Supplier Meso Scale Discovery Supplier No. K251VAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MIP-3beta 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer It can test up to 80 samples in duplicate and has a testing time of 3 5 hours