Learn More
Meso Scale Discovery UPLEX HUM MMP1 ASY384well
SDP
Catalog No. 5000080814
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The lower limit of detection is 0 03 ng/mL with an upper limit of 10 000 ng/mL The assay measures active MMP-1 and is supplied with plates detection antibodies calibrators and ancillary reagents for performing the immunoassay
Catalog No. 50-000-80814 Supplier Meso Scale Discovery Supplier No. K251AHDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of MMP-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The lower limit of detection is 0 03 ng/mL with an upper limit of 10 000 ng/mL The assay measures active MMP-1 and is supplied with plates detection antibodies calibrators and ancillary reagents for performing the immunoassay