Learn More
Meso Scale Discovery UPLEX HUM MMP1 ASYQP 25 PL
SDP
Catalog No. 5000081659
Encompass
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay that measures MMP-1 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can be multiplexed to measure up to 10 biomarkers per well It takes approximately 3 5 hours to complete and includes reagents for 96 tests in the kit The U-PLEX MMP-1 assay delivers rapid precise quantification of MMP-1 levels
Catalog No. 50-000-81659 Supplier Meso Scale Discovery Supplier No. K151AHDK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-1 Assay is a multiplex immunoassay that measures MMP-1 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can be multiplexed to measure up to 10 biomarkers per well It takes approximately 3 5 hours to complete and includes reagents for 96 tests in the kit The U-PLEX MMP-1 assay delivers rapid precise quantification of MMP-1 levels