Learn More
Meso Scale Discovery UPLEX HUM MMP2 ASY384well
SDP
Catalog No. 5000080749
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a 384-well multiplex immunoassay kit that measures active and pro-MMP-2 levels in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for MMP-2
Catalog No. 50-000-80749 Supplier Meso Scale Discovery Supplier No. K251AHKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a 384-well multiplex immunoassay kit that measures active and pro-MMP-2 levels in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for MMP-2