Learn More
Meso Scale Discovery UPLEX HUM MMP2 ASYQP 25 PL
SDP
Catalog No. 5000078401
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-2 in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MMP-2 in samples
Catalog No. 50-000-78401 Supplier Meso Scale Discovery Supplier No. K151AHKK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-2 in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MMP-2 in samples