Learn More
Meso Scale Discovery UPLEX HUM MMP7 ASY384well
SDP
Catalog No. 5000081135
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay has high sensitivity detecting MMP-7 concentrations as low as 0 03 ng/mL and a broad dynamic range of up to 10 000 pg/mL
Catalog No. 50-000-81135 Supplier Meso Scale Discovery Supplier No. K251AHEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay has high sensitivity detecting MMP-7 concentrations as low as 0 03 ng/mL and a broad dynamic range of up to 10 000 pg/mL