Learn More
Meso Scale Discovery UPLEX HUM MMP7 ASYQP 25 PL
SDP
Catalog No. 5000080490
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrator solutions and sample diluent It provides a simple reproducible method to quantify MMP-7 levels for research use
Catalog No. 50-000-80490 Supplier Meso Scale Discovery Supplier No. K151AHEK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrator solutions and sample diluent It provides a simple reproducible method to quantify MMP-7 levels for research use