Learn More
Meso Scale Discovery UPLEX HUM MMP9 total ASYQP
SDP
Catalog No. 5000079557
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels from small sample volumes This assay has high specificity and no cross-reactivity with other MMP family members It requires an MSD instrument such as QuickPlex SQ120 or SECTOR Imager for detection The kit provides reagents for 96 tests in a quick 4 5 hour protocol
Catalog No. 50-000-79557 Supplier Meso Scale Discovery Supplier No. K151AHMK22
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels from small sample volumes This assay has high specificity and no cross-reactivity with other MMP family members It requires an MSD instrument such as QuickPlex SQ120 or SECTOR Imager for detection The kit provides reagents for 96 tests in a quick 4 5 hour protocol