Learn More
Meso Scale Discovery UPLEX HUM MMP9 TTL ASY384well
SDP
Catalog No. 5000080819
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels with high specificity and low background This assay kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and buffer for sample dilution It allows high-throughput analysis of MMP-9 levels with a minimal sample volume of 25 L per well
Catalog No. 50-000-80819 Supplier Meso Scale Discovery Supplier No. K251AHMK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-9 levels with high specificity and low background This assay kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and buffer for sample dilution It allows high-throughput analysis of MMP-9 levels with a minimal sample volume of 25 L per well