Learn More
Meso Scale Discovery UPLEX HUM MMP9 TTL ASY384well
SDP
Catalog No. 5000080991
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is a 384-well electrochemiluminescent immunoassay designed to measure total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of MMP-9 levels with as little as 25 L sample volume per well The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of quantification of 10 000 pg/mL It includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator containing recombinant MMP-9
Catalog No. 50-000-80991 Supplier Meso Scale Discovery Supplier No. K251AHMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is a 384-well electrochemiluminescent immunoassay designed to measure total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of MMP-9 levels with as little as 25 L sample volume per well The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of quantification of 10 000 pg/mL It includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator containing recombinant MMP-9