Learn More
Meso Scale Discovery UPLEX HUM MMP9 TTL ASYQP
SDP
Catalog No. 5000081653
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in just 3 5 hours The assay has a detection range of 0 03-1500 ng/mL and includes a 96-well plate pre-coated with capture antibody detection antibody calibrator and sample diluent This assay allows for the accurate quantification of total MMP-9 levels in various human sample types
Catalog No. 50-000-81653 Supplier Meso Scale Discovery Supplier No. K151AHMK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in just 3 5 hours The assay has a detection range of 0 03-1500 ng/mL and includes a 96-well plate pre-coated with capture antibody detection antibody calibrator and sample diluent This assay allows for the accurate quantification of total MMP-9 levels in various human sample types