Learn More
Meso Scale Discovery UPLEX HUM Nectin4 ASY 25 PL
SDP
Catalog No. 5000078406
Encompass
The Meso Scale Discovery U-PLEX Human Nectin-4 Assay is a multiplex immunoassay kit that enables simultaneous quantification of Nectin-4 in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This product allows for the measurement of Nectin-4 which is a biomarker associated with various cancers With minimal sample volume requirements this assay kit is well-suited for cancer research and drug development applications
Catalog No. 50-000-78406 Supplier Meso Scale Discovery Supplier No. K151AHFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Nectin-4 Assay is a multiplex immunoassay kit that enables simultaneous quantification of Nectin-4 in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This product allows for the measurement of Nectin-4 which is a biomarker associated with various cancers With minimal sample volume requirements this assay kit is well-suited for cancer research and drug development applications