Learn More
Meso Scale Discovery UPLEX HUM Nectin4 ASY384well
SDP
Catalog No. 5000080747
Encompass
The Meso Scale Discovery Nectin-4 Assay Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative detection of human Nectin-4 in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a rapid accurate and robust assay with high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with a monoclonal antibody to human Nectin-4 a SULFO-TAG labeled detection antibody assay and sample diluents calibrators and a plate reader The assay has a typical sensitivity of 0 3 ng/mL and a dynamic range of up to 10 000 pg/mL
Catalog No. 50-000-80747 Supplier Meso Scale Discovery Supplier No. K251AHFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Nectin-4 Assay Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative detection of human Nectin-4 in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a rapid accurate and robust assay with high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with a monoclonal antibody to human Nectin-4 a SULFO-TAG labeled detection antibody assay and sample diluents calibrators and a plate reader The assay has a typical sensitivity of 0 3 ng/mL and a dynamic range of up to 10 000 pg/mL