Learn More
Meso Scale Discovery UPLEX HUM NGAL/LCN2 ASY384
SDP
Catalog No. 5000079478
Encompass
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) 384-well assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of NGAL in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening This assay measures NGAL which is involved in innate immunity to bacterial infections It can aid in the risk assessment of acute kidney injury This assay takes approximately 4 hours to run and has a detection range of 45 3-30000 pg/mL
Catalog No. 50-000-79478 Supplier Meso Scale Discovery Supplier No. K251Z1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) 384-well assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of NGAL in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening This assay measures NGAL which is involved in innate immunity to bacterial infections It can aid in the risk assessment of acute kidney injury This assay takes approximately 4 hours to run and has a detection range of 45 3-30000 pg/mL