Learn More
Meso Scale Discovery UPLEX HUM NGAL/LCN2 ASY384well
SDP
Catalog No. 5000080564
Encompass
The Meso Scale Discovery U-PLEX Human NGAL 384-Well Assay is a multiplex immunoassay kit for measuring NGAL in human serum plasma urine saliva CSF and cell culture supernatants using electrochemiluminescence detection It uses MSD s SULFO-TAG labels offering a sensitive accurate and reproducible method with a broad dynamic range The assay requires 50 L of sample and completes in under 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrator solutions and necessary buffers
Catalog No. 50-000-80564 Supplier Meso Scale Discovery Supplier No. K251Z1K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human NGAL 384-Well Assay is a multiplex immunoassay kit for measuring NGAL in human serum plasma urine saliva CSF and cell culture supernatants using electrochemiluminescence detection It uses MSD s SULFO-TAG labels offering a sensitive accurate and reproducible method with a broad dynamic range The assay requires 50 L of sample and completes in under 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrator solutions and necessary buffers