Learn More
Meso Scale Discovery UPLEX HUM NGAL/LCN2 ASYQP
SDP
Catalog No. 5000080487
Encompass
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in human serum plasma and urine This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other human proteins It requires a low sample volume of 50 L and has high reproducibility with intra- and inter-assay %CVs of 10% The U-PLEX Human NGAL Assay provides reliable sensitive and rapid quantification of NGAL an early biomarker of acute kidney injury
Catalog No. 50-000-80487 Supplier Meso Scale Discovery Supplier No. K151Z1K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in human serum plasma and urine This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other human proteins It requires a low sample volume of 50 L and has high reproducibility with intra- and inter-assay %CVs of 10% The U-PLEX Human NGAL Assay provides reliable sensitive and rapid quantification of NGAL an early biomarker of acute kidney injury