Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASY
SDP
Catalog No. 5000078518
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay that measures OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents needed to perform the assay It can accurately quantify OX40 levels with minimal hands-on time using an MSD instrument
Catalog No. 50-000-78518 Supplier Meso Scale Discovery Supplier No. K151T7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay that measures OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents needed to perform the assay It can accurately quantify OX40 levels with minimal hands-on time using an MSD instrument