Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASY384
SDP
Catalog No. 5000079409
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of OX40 and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires a low sample volume of 25 L and the total assay time is less than 3 5 hours The kit includes a 10-plate kit with reagents and plates sufficient for 3 840 data points when used as directed This assay allows for efficient and reproducible analysis of OX40 and other immune checkpoint biomarkers
Catalog No. 50-000-79409 Supplier Meso Scale Discovery Supplier No. K251T7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) 384-well Assay is a multiplex immunoassay kit that enables simultaneous quantification of OX40 and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires a low sample volume of 25 L and the total assay time is less than 3 5 hours The kit includes a 10-plate kit with reagents and plates sufficient for 3 840 data points when used as directed This assay allows for efficient and reproducible analysis of OX40 and other immune checkpoint biomarkers