Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASYQP
SDP
Catalog No. 5000082305
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay that measures OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents needed to perform the assay It can detect OX40 levels as low as 17 pg/mL with an upper limit of 100 000 pg/mL The assay takes 3 5 hours to complete
Catalog No. 50-000-82305 Supplier Meso Scale Discovery Supplier No. K151T7K21
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay that measures OX40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluents needed to perform the assay It can detect OX40 levels as low as 17 pg/mL with an upper limit of 100 000 pg/mL The assay takes 3 5 hours to complete