Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 1 ASY
SDP
Supplier:  Meso Scale Discovery K151U7K1
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit (K151U7K) is a multiplex immunoassay that measures PD1 Epitope 1 in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent Results can be read on MSD SECTOR Imager plate readers This assay allows for the quantitative measurement of PD1 Epitope 1 which may have utility in cancer immunotherapy research and drug development
Catalog No. 50-000-79001