Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 1 ASY
SDP
Catalog No. 5000080404
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit is a multiplex immunoassay that measures PD1 Epitope 1 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to quantify PD1 Epitope 1 levels This assay enables the quantification of PD1 Epitope 1 levels which can provide insights into immune checkpoint inhibitor therapy response
Catalog No. 50-000-80404 Supplier Meso Scale Discovery Supplier No. K151U7K2
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit is a multiplex immunoassay that measures PD1 Epitope 1 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to quantify PD1 Epitope 1 levels This assay enables the quantification of PD1 Epitope 1 levels which can provide insights into immune checkpoint inhibitor therapy response