Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 1 ASYQP
SDP
Catalog No. 5000079117
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit (K151U7K) is a multiplex immunoassay for measuring PD1 Epitope 1 in human serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls for a standard curve It quantitatively measures PD1 Epitope 1 with accuracy and precision aiding the study of immune checkpoint pathways
Catalog No. 50-000-79117 Supplier Meso Scale Discovery Supplier No. K151U7K22
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit (K151U7K) is a multiplex immunoassay for measuring PD1 Epitope 1 in human serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls for a standard curve It quantitatively measures PD1 Epitope 1 with accuracy and precision aiding the study of immune checkpoint pathways