Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASY
SDP
Catalog No. 5000078302
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD1 epitope 2 and total PD1 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay needs 50 L of sample and is read on MSD s SECTOR Imager 6000 The kit includes necessary components for generating a standard curve and validating assay performance enabling monitoring of PD1 epitope occupancy in samples from subjects treated with anti-PD1 therapeutics
Catalog No. 50-000-78302 Supplier Meso Scale Discovery Supplier No. K151V7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD1 epitope 2 and total PD1 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay needs 50 L of sample and is read on MSD s SECTOR Imager 6000 The kit includes necessary components for generating a standard curve and validating assay performance enabling monitoring of PD1 epitope occupancy in samples from subjects treated with anti-PD1 therapeutics