Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASY
SDP
Catalog No. 5000079682
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous measurement of PD1 epitope 2 biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It can be used for biomarker discovery lead optimization and immunogenicity testing With a short 3 5 hour protocol it delivers rapid results using small sample volumes
Catalog No. 50-000-79682 Supplier Meso Scale Discovery Supplier No. K151V7K2
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous measurement of PD1 epitope 2 biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It can be used for biomarker discovery lead optimization and immunogenicity testing With a short 3 5 hour protocol it delivers rapid results using small sample volumes