Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASY
SDP
Catalog No. 5000082039
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD1 epitope 2 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and compatibility with small sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and measuring PD1 epitope 2 concentrations aiding immunotherapy research
Catalog No. 50-000-82039 Supplier Meso Scale Discovery Supplier No. K151V7K1
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD1 epitope 2 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and compatibility with small sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve and measuring PD1 epitope 2 concentrations aiding immunotherapy research